Utility wipes, WYPALL* L10
Supplier: KIMBERLY CLARK
05322
KIMB05322EA 408 SGD
KIMB05322
Utility wipes, WYPALL* L10
Wipes Disposable Wipes and Towels Wipes for General Applications
These paper wipes are perfect for spray and wipe jobs, glass surface cleaning, absorbing small liquid spills, detail and final assembly wiping.
  • One-ply wipes
  • Strong, soft, highly absorbent, lint-free and odorless
  • Specially treated for wet strength
  • Meet EPA guidelines for recycled fibre content
Colour: White