Wipes for WETTASK™ system
Supplier: KIMBERLY CLARK
06001
KIMB06001EA 220 SGD
KIMB06001
Wipes for WETTASK™ system
Wipes Disposable Wipes and Towels Presaturated Wipes
The Kimtech™ Prep Wipers for the WetTask System, for Solvents, is built for critical tasks, such as preparing cleaning surfaces.
  • Cuts solvent usage by 20% using the Wettask closed bucket
  • Conveniently portable
  • Eliminate the need for laundering contaminated cloth rags
The enclosed system also helps eliminate exposure to chemical vapors and splashes. The Wettask closed bucket and wet wiping system can rreduce VOC emissions for a safer, more cost effective workplace.
Packaging: 6 rolls / case, 95 sheets / roll, 570 sheets / case.
Order Now
SPECIFICATIONS
Environmentally Preferable