Cleanroom wipes, pre-saturated, alcohol, Kimtech™ W4
Supplier: KIMBERLY CLARK
06070
KIMB06070EA 316 SGD
KIMB06070
Cleanroom wipes, pre-saturated, alcohol, Kimtech™ W4
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
The 100% polypropylene wipers come pre-saturated with 70% isopropyl alcohol (semiconductor grade) and 30% deionized water, in a resealable pouch.
  • Ideal one-step application of alcohol to critical surfaces
  • Low lint for environments where 'lint free' standards are important
  • These disposable wipers are recommended for ISO Class 4 and higher clean rooms
These disposable wipers are ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries that require alcohol wiping for critical tasks.
Certifications: Meets EPA guidelinesMore Product Information