Cleanroom wipes, Kimtech™ Pure W5
Supplier: KIMBERLY CLARK
06179
KIMB06179EA 27.4 SGD
KIMB06179
Cleanroom wipes, Kimtech™ Pure W5
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These critical task wipes feature a 50% rayon/50% polyester spunlace blend base sheet with high water absorbency rate, plus excellent absorption efficiency and capacity.
  • No binders or surfactants are used resulting in low extractables
  • Clothlike feel
  • Double bagged with case liner
Suitable for use in Class 100, ISO 5, M2.5 or higher cleanrooms.
Colour: white
Packaging: Double-bagged in anti-stat packaging.