Safety goggles, JACKSON SAFETY™ V80
Supplier: KIMBERLY CLARK
16361
KIMB16361EA 17.6 SGD
KIMB16361
Safety goggles, JACKSON SAFETY™ V80
Goggles
KleenGuard™ Personal Protective Equipment (PPE) enables allowing for maximum productivity and safety for you and your employees.
  • Splash protection
  • Larger lens for enhanced fit and comfort
  • A wider nose piece to fit over most half mask respirators
  • Provide 99.9% UVA/UVB/UVC protection
  • Polycarbonate lenses are hard coated for scratch resistance
Each pair features indirect ventilation along with two additional baffled ports for increased airflow and minimized fogging and condensation. The headband buckles are located toward the rear of the goggles to limitMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable