Visitor safety spectacles, KleenGuard™ Unispec™ II
Supplier: KIMBERLY CLARK
16727
KIMB16727EA 0 SGD
KIMB16727
Visitor safety spectacles, KleenGuard™ Unispec™ II
Safety Glasses
These spectacles offer wraparound protection to fit over most prescription eyewear, in a metal-free design with screw-less hinges.
  • Hard coated lenses for scratch resistance and longer life
  • Polycarbonate lenses provide 99,9% UVA/UVB/UVC protection
This clear safety glasses (with clear temples) allow maximum visible light transmission, which is ideal for indoor use.
Certifications: Meet the ANSI Z87.1+ standards for impact and are TAA compliant.
Packaging: 50 pairs safety spectacles /case.
Order Now
SPECIFICATIONS
Environmentally Preferable