Safety spectacles, V60 Nemesis* RX, JACKSON SAFETY*
Supplier: KIMBERLY CLARK
28630
KIMB28630EA 0 SGD
KIMB28630
Safety spectacles, V60 Nemesis* RX, JACKSON SAFETY*
Safety Glasses
Lightweight safety spectacles with a wrap-around design for added comfort and coverage.
  • 99,9% UVA/UVB protection
  • Bifocal style with diopters for vision assistance
  • Enhanced nose piece design to channel away sweat
  • Every pair includes a neck cord
The Nemesis™ Rx readers re designed for long-term comfort, thanks to flexible and lightweight design, soft touch temples, and a nosepiece with integrated ridges to channel away sweat. These safety glasses have polycarbonate lenses that provide 99,9% UVA/UVB/UVC protection to help prevent eye damage likeMore Product Information