Kimtech Science™ Kimwipes™ Wipes for Delicate Tasks
Supplier: KIMBERLY CLARK
341550LS
KIMB341550LSEA 358 SGD
KIMB341550LS
Kimtech Science™ Kimwipes™ Wipes for Delicate Tasks
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
These light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
  • Anti-stat dispensing reduces lint and electrostatic discharge and controls usage
  • Portability reduces waste
  • One-at-a-time dispensing (Pop-Up™ Box dispenser)
  • Also available in 2-ply and 3-ply for increased absorbency
  • Gentle texture
Used in environments like research facilities, pharmaceutical, biotech, high-tech, manufacturing.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904