Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Supplier: KIMBERLY CLARK
21903-021 34705
KIMB21903-021EA 640.86 SGD
KIMB21903-021 KIMB34705
Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
This 2-ply wipe offers a unique combination of low-lint tissue softness and multi-ply absorbency. Perfect for a multitude of delicate tasks.
  • Multi-ply absorbency wipes up liquid and dust
  • Wipes clean with extra low-lint and low extractable performance
  • One-at-a-time dispensing