Cleanroom sleeve protectors, Kimtech™ A5
Supplier: KIMBERLY CLARK
36077
KIMB36077EA 391 SGD
KIMB36077
Cleanroom sleeve protectors, Kimtech™ A5
Disposable Garments
These sterile sleeve protectors are made of durable SMS fabric which provides a cloth-like feel. A highly visible blue line along the inside of the sleeve signals the proper place to grasp while donning, helping workers avoid touching the outside.
  • Thumb loop keeps sleeve from rolling back and exposing arm or wrist
  • Tunnelled elastic and bound seams
  • Triple-bagged
Suitable for Class 100, ISO Class 5 or higher cleanrooms.
Certifications: PPE Cat I (EU) 2016/425.