Wipes for general applications, WYPALL* L20
Supplier: KIMBERLY CLARK
47044-50
KIMB47044-50EA 25 SGD
KIMB47044-50
Wipes for general applications, WYPALL* L20
Wipes Disposable Wipes and Towels Wipes for General Applications
These high-capacity, single-use paper wipers are excellent for Large spills. They are also ideal for parts and tool cleanups, mining operations maintenance, hydraulic leak absorption, general-purpose field utility wiping, conveyor and roller cleaning, and retail store maintenance and cleaning.
  • Textured side for better cleaning power
  • Use with dispenser for neat and controlled dispensing
  • Practical Pop-Up* box for easy dispensing
Order Now
SPECIFICATIONS
Environmentally Preferable