Kimtech™ Polaris™ Nitrile Gloves
Supplier: KIMBERLY CLARK
62000 62001 62002 62003 62004
KIMB62000EA 25.8 SGD
KIMB62000 KIMB62001 KIMB62002 KIMB62003 KIMB62004
Kimtech™ Polaris™ Nitrile Gloves
Gloves
Kimtech™ Polaris™ Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments.
  • High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
  • Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
  • Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
  • Extensively tested
More Product Information