Nitrile gloves, Kimtech™ Polaris™ Xtra
Supplier: KIMBERLY CLARK
62100 62101 62102 62103 62104
KIMB62100EA 17.8 SGD
KIMB62100 KIMB62101 KIMB62102 KIMB62103 KIMB62104
Nitrile gloves, Kimtech™ Polaris™ Xtra
Gloves
Kimtech™ Polaris™ Xtra Nitrile Gloves are built for science, made and designed for use in laboratories, life science applications, and non-sterile pharmaceutical manufacturing environments.
  • High-end ambidextrous powder-free nitrile gloves deliver seamless protection whenever and wherever it is needed
  • Optimized nitrile formulation (acrylonitrile-butadiene) features extended chemicals and chemotherapy drugs splash protection without compromising comfort
  • Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials
  • Extensively
More Product Information