Cleanroom face masks, Kimtech™ M3, PE/BiCoSof™
Supplier: KIMBERLY CLARK
62484 62465 62466 62466
KIMB62484EA 149 SGD
KIMB62484 KIMB62465 KIMB62466 TECO62466
Cleanroom face masks, Kimtech™ M3, PE/BiCoSof™
Masks Cleanroom Masks
The pleat-style masks have soft ties and a fully enclosed bendable nosepiece.
  • Outer layer of porous PE film protects from contamination
  • Excellent breathability with filtration of ≥99% at 0,5 µm
  • Inner facing made of white BiCoSof™ and fully enclosed, soft, malleable aluminium nosepiece
Suitable for Class 10, ISO 4, M2.5 cleanrooms.
Packaging: 50 per bag, 6 or 10 bags per case.