Disposable particle filtering respirators, FFP1/FFP2/FFP3
Supplier: KIMBERLY CLARK
64230
KIMB64230EA 0 SGD
KIMB64230
Disposable particle filtering respirators, FFP1/FFP2/FFP3
Respirators Disposable Respirators Particulate Respirators
Moulded masks made from three-layer filter media. Silicone and latex-free.
  • Adjustable comfort head straps increase comfort and ensure secure fit
  • Colour coding for easy recognition of performance level
  • Nose seal is made from water repellent material for better sealing, reduces fogging on safety glasses
  • Tested at 120 mg, increases capacity to operate longer in dusty environments
Certifications: EN 149:2001
Order Now
SPECIFICATIONS
Environmentally Preferable