Wipes and wipe rolls for general applications, WYPALL* L20 EXTRA / L20 EXTRA+
Supplier: KIMBERLY CLARK
7300 7301 7303 7302
KIMB7300EA 58.4 SGD
KIMB7300 KIMB7301 KIMB7303 KIMB7302
Wipes and wipe rolls for general applications, WYPALL* L20 EXTRA / L20 EXTRA+
Wipes Disposable Wipes and Towels Wipes for General Applications
These single-use wipes, made from two ply AIRFLEX* material, are ideal for multipurpose cleaning and medium duty wiping tasks such as polishing glass, smooth surfaces and cleaning tools.
  • Stronger than conventional two ply wipes
  • Strong and absorbent
  • Clean up fast without disintegrating
  • Can be used with a dispensing system to help control use and reduce wastage
Small rolls: For use with dispenser 7041.
Large rolls: For use with dispensers 6146, 6154 and 6155.
Centrefeed rolls: For use with dispenser 7017.