Wipes rolls, WYPALL* L30 ULTRA+
Supplier: KIMBERLY CLARK
7425 7426
KIMB7425EA 175 SGD
KIMB7425 KIMB7426
Wipes rolls, WYPALL* L30 ULTRA+
Wipes Disposable Wipes and Towels Wipes for General Applications
These single use wipes made from three ply AIRFLEX* material are ideal for superior cleaning and heavy duty wiping tasks such as soaking up industrial spills, cleaning parts and components.
  • Strongest, thickest and most absorbent WYPALL L wipe
  • Stronger than conventional three ply wipes
  • Available as a white or blue perforated large roll
  • Can be used with a dispensing system to help control use and reduce wastage
For use with dispensers 115-0045, 115-1533 and 115-2038.
Order Now
SPECIFICATIONS
Environmentally Preferable