Wipes for general applications, WYPALL* L40
Supplier: KIMBERLY CLARK
7462
KIMB7462EA 67.5 SGD
KIMB7462
Wipes for general applications, WYPALL* L40
Wipes Disposable Wipes and Towels Wipes for General Applications
WYPALL* L40 wipes easily absorb large spills and leaks and are therefore ideal for routine industrial cleaning and maintenance. They are also soft enough to remove soils and liquids from face and hands.
  • Ideal for benchtop use
  • Made of a cellulose/latex mixture
Colour: White.