Surface preparation cloth, KIMTECH PREP*
Supplier: KIMBERLY CLARK
7589
KIMB7589EA 0 SGD
KIMB7589
Surface preparation cloth, KIMTECH PREP*
Wipes Reusable Cleaning Cloths Cloths for General Applications
This highly absorbent multipurpose cleaning and polishing cloth made from microfibre can be used wet or dry. It is effective in trapping and removing particulate matter from a variety of surfaces.
  • Low linting which reduces the risk of process contamination
  • Machine washable up to 300 times
  • Soft and flexible
  • Silicone-free, seamed edges