Cleanroom wipes, Kimtech™ Pure W4
Supplier: KIMBERLY CLARK
7605
KIMB7605EA 503 SGD
KIMB7605
Cleanroom wipes, Kimtech™ Pure W4
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipes made from 100% PP are ideal for special cleaning applications in critical areas thanks to their thermal bonding (i.e. no adhesive or binding agent).
  • Delivered double-packed
  • Cut edges
  • Resistant to acids, alkalis and solvents
Suitable for use in Class 10, ISO 4, M2.5 cleanrooms.
Colour: White