Cleanroom wipes, pre-saturated, sterile, KIMTECH PURE* W4
Supplier: KIMBERLY CLARK
76490
KIMB76490EA 285 SGD
KIMB76490
Cleanroom wipes, pre-saturated, sterile, KIMTECH PURE* W4
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
This sterile non woven PP wipe is pre-saturated with 70% isopropyl alcohol and 30% deionized water. Recommended for the most critical wiping tasks in ISO Class 4 or higher cleanroom environments.
  • Ideal one-step application of alcohol to critical surfaces
  • Resealable pouch with case liner
Suitable for use in class 10, ISO 4, M2.5 cleanrooms.
Order Now
SPECIFICATIONS
Environmentally Preferable