WypAll® X50 Wipers
Supplier: KIMBERLY CLARK
83550
KIMB83550EA 231 SGD
KIMB83550
WypAll® X50 Wipers
Wipes Reusable Cleaning Cloths Cloths for General Applications
Strong and sensible, WYPALL® X50 Wipers offer economical, extended-use wiper performance in a variety of applications.
WYPALL® X50 Wipers combine versatile performance with economical design for light-duty tasks. Ideal for manufacturing, healthcare, and transportation environments. Reinforced for strength when wet or dry. HYDROKNIT™ Fabric eliminates adhesives and binders, and eliminates left-behind residue.