Cleanroom overalls, Kimtech™ A5
Supplier: KIMBERLY CLARK
88800 88801 88802 88803 88804
KIMB88800EA 391 SGD
KIMB88800 KIMB88801 KIMB88802 KIMB88803 KIMB88804
Cleanroom overalls, Kimtech™ A5
Disposable Garments
Kimtech™ A5 sterile cleanroom apparel with CLEAN-DON™ technology is made of SMS (Spunbond Melt-blown Spunbond) material. It features built-in snaps that gather up legs and arms to aid aseptic gowning. Garments have a thumb loop that prevents the sleeve from rolling back and exposing arm or wrist when in use.
  • Cloth-like comfort
  • Innovative inside-out fold pattern - reduces the risk of touching and contaminating the outside of the apparel when opening or donning
  • Highly visible blue line along the inside of the garment signals the proper place to grasp while
More Product Information