Cleanroom gloves, nitrile, Kimtech™ G3 and G5 Sterling™
Supplier: KIMBERLY CLARK
98186_U
KIMB98186_UEA 0 SGD
KIMB98186_U
Cleanroom gloves, nitrile, Kimtech™ G3 and G5 Sterling™
Gloves
Ambidextrous latex-free gloves with beaded cuffs.
  • Non sterile, powder-free
  • 100% nitrile for lower risk of glove-induced allergic type 1 reactions
  • Textured fingertips for improved grip
  • Static dissipative material for reduced ESD effect
  • Sterling™ process allows a reduction in glove thickness, so less force is required to stretch the material
  • Reduced glove thickness requires less raw materials in production which significantly reduces waste and saves storage space
Typical applications include: Semiconductor, medical device, flat panel display,More Product Information