Milk dilution bottles, KIMBLE®
Supplier: KIMBLEKONTES
14915-160 14925-160
KIML14915-160EA 638 SGD
KIML14915-160 KIML14925-160
Milk dilution bottles, KIMBLE®
Bottles Dilution Bottles
Manufactured from USP Type 1 borosilicate molded glass.
  • Autoclavable to 121 °C without preconditioning
  • Black phenolic screw cap, supplied unattached, has a cemented-in rubber liner and is suitable for autoclaving
  • Bottles have a square cross-section
These KIMAX®-35 bottles are designed from the requirements for milk dilution bottles given in the 'Standard Methods for the Examination of Dairy Products,' published by the American Public Health Association.