Dropping bottles, glass, with taper pipette, KIMBLE®
Supplier: KIMBLEKONTES
15035-125
KIML15035-125EA 1690 SGD
KIML15035-125
Dropping bottles, glass, with taper pipette, KIMBLE®
Bottles Dropping Bottles
The dropper bottle, manufactured from USP Type 1 borosilicate molded glass, is for general purpose use and for determining the oil absorption of pigments (ASTM D281).
  • Excellent temperature resistance
  • Excellent chemical resistance
Delivery information: Available with a medium length standard taper pipet dropper and a rubber nipple.