Specific gravity bottles, Hubbard, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
15110-24
KIML15110-24EA 2650 SGD
KIML15110-24
Specific gravity bottles, Hubbard, KIMAX®, KIMBLE®
Specific Gravity Bottles
Borosilicate glass, KIMAX®.
Bottle used in the determination of specific gravity for a large variety of semi-solid materials and emulsions according to ASTM D70, D1429, D115. It features short length conical stopper on the underside and has a hole approximately 1,6 mm in Ø to allow air to escape.
Certifications: KIMAX® glass conforms to USP Type I and ASTM E438, Type I, class A requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable