Weighing bottles, glass, KIMBLE®
Supplier: KIMBLEKONTES
15157-2024
KIML15157-2024EA 750 SGD
KIML15157-2024
Weighing bottles, glass, KIMBLE®
Bottles Weighing Bottles
Manufactured from 33 expansion, low extractable borosilicate.
  • Excellent temperature resistance
  • Excellent chemical resistance
These weighing bottles are designed for weighing small samples of coal.The cap-style closure fits over externally ground body.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements