Pressure release valves, AIRLESS-WARE®, KIMBLE®
Supplier: KIMBLEKONTES
216100-0000
KIML216100-0000EA 635 SGD
KIML216100-0000
Pressure release valves, AIRLESS-WARE®, KIMBLE®
Valves Pressure-Relief Valves
Designed for applications that require evacuated apparatus to be filled with an inert gas to one atmosphere.Overall height 130 mm, Overall depth 102 mm, overall width 80 mm. It fits a tubing of internal diameter ³/₈ inches.
  • Uses include the introduction of an inert gas into a vacuum desiccator or nitrogen into Airless-ware™ apparatus
  • A pressure release valve should be placed between the nitrogen source and nitrogen manifold on all Airless-ware™ manifold installations
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type
More Product Information