Petri dishes, KIMBLE® KIMAX®
Supplier: KIMBLEKONTES
23060-15020
KIML23060-15020EA 906 SGD
KIML23060-15020
Petri dishes, KIMBLE® KIMAX®
Culture Dishes Petri Dishes
KIMAX®, borosilicate glass Petri dishes with reinforced beaded edges to resist mechanical breakage and to help in centring the bottom inside the cover.
  • Dishes remain clear after repeated use in wet or dry sterilisation cycles
  • Covers have a fused-on red ceramic enamel marking spot and inscription, including 'Cover'
  • Bottoms have a white ceramic enamal inscription 'Bottom'
  • Bottom has a vertical arrow on the side to precisely locate serial dilution starting points
  • Combination of different colours and wording permits quick and easy identification of parts when
More Product Information