Soxhlet extractor, KIMBLE®
Supplier: KIMBLEKONTES
24005-50
KIML24005-50EA 1120 SGD
KIML24005-50
Soxhlet extractor, KIMBLE®
Extractors Soxhlet and Soxhlet-Type Extractors
Borosilicate glass.
  • Approximate capacities are measured to the base of the flask neck
  • Standard joint size between Soxhlet tube and condenser is 34/45 and between the Soxhlet tube and flask is 24/40
  • Thimble size is determined by the ID of the extraction tube
  • Hose connections on condenser accept ⁵/₁₆" int.Ø flexible tubing
The Soxhlet extractor was designed for separating an analyte with limited solubility in a particular solvent from an insoluble compound. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.