Erlenmeyer flasks, baffled, KIMBLE®
Supplier: KIMBLEKONTES
25630-125
KIML25630-125EA 303 SGD
KIML25630-125
Erlenmeyer flasks, baffled, KIMBLE®
Flasks Erlenmeyer Flasks
Erlenmeyer flasks, manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • With white ceramic enamel marking spots
Designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to improve oxygen transfer rates or aeration. And also this flask have a neck with a plain fire-polished finishMore Product Information