Erlenmeyer flasks, narrow neck, reinforced beaded top, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
26500-125 26500-6000 26500-2000 26500-300 26500-50 26500-250 26500-1000 26520-1
KIML26500-125EA 448 SGD
KIML26500-125 KIML26500-6000 KIML26500-2000 KIML26500-300 KIML26500-50 KIML26500-250 KIML26500-1000 KIML26520-1
Erlenmeyer flasks, narrow neck, reinforced beaded top, KIMAX®, KIMBLE®
Flasks Erlenmeyer Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Durable white ceramic enamel scales to indicate approximate volumes at various levels
  • Good chemical resistance
  • Good temperature resistance
Versatile flasks for general laboratory use. Tops are reinforced and tooled for maximum mechanical strength. Body has thick walls with a long tapered outside contour to minimize chipping.
Certifications: Designed from ASTM Specification E1404, Type I, Class I requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class AMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable