Erlenmeyer flasks, with pennyhead glass stopper, KIMBLE®
Supplier: KIMBLEKONTES
26600-250 26600-125 26600-500
KIML26600-250EA 885 SGD
KIML26600-250 KIML26600-125 KIML26600-500
Erlenmeyer flasks, with pennyhead glass stopper, KIMBLE®
Flasks Erlenmeyer Flasks
Manufactured from 33 expansion, KIMAX®, low extractable borosilicate glass.
  • With a standard taper ground glass stopper neck finish
  • With capacity scale marking
  • Available with stopper
Designed from ASTM Specification E1404, Type II, Class II requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.