Volumetric flasks, class A, glass, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
28046-900
KIML28046-900EA 449 SGD
KIML28046-900
Volumetric flasks, class A, glass, KIMAX®, KIMBLE®
Flasks Volumetric Flasks
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Calibrated both 'to contain' and 'to deliver' (top line)
  • Save shelf space, requires less space than a round flask (25 to 30%)
  • Graduation rings and marking spot are blasted onto glass
These square volumetric flasks are designed to take up less area and pack more closely on shelves, in refrigerators and in other storage cabinets.
The flasks have a square cross-section with a width less than the widest diameter of a typical round flask. Due to the added glass weight and the squareMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable