Volumetric flasks, class A, cassia style, KIMBLE®
Supplier: KIMBLEKONTES
28066-110
KIML28066-110EA 1020 SGD
KIML28066-110
Volumetric flasks, class A, cassia style, KIMBLE®
Flasks Volumetric Flasks
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®
  • Calibrated to contain
  • With a marking spot and a durable white ceramic enamel scale
This flask is used in the assay of oils of cinnamon, chenopodium, cloves, spearmint and other essential oils.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.