Gooch crucibles, with fritted disc, KIMFLOW®, KIMBLE®
Supplier: KIMBLEKONTES
28260-301
KIML28260-301EA 925 SGD
KIML28260-301
Gooch crucibles, with fritted disc, KIMFLOW®, KIMBLE®
Crucibles
These Gooch crucibles are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Designed for collecting, drying and weighing precipitates and samples to be analysed using gravimetric analysis. These crucibles are suitable for precipitates to be dried to a constant weight at 110 °C.
The fritted disc is sealed into the crucible.
At higher temperatures, heating in an electric furnace is advised, and exposure to sudden temperature changes should be avoided. For safe handling, crucibles should not beMore Product Information