Funnels, with long stem, KIMBLE®
Supplier: KIMBLEKONTES
28900-75
KIML28900-75EA 1130 SGD
KIML28900-75
Funnels, with long stem, KIMBLE®
Funnels Filtration Funnels Filter Funnels
KIMAX®, borosilicate glass, with 58° cone angle.
  • High resistance to chemical attack
  • Resistant to mechanical and thermal shock
  • ASTM Specification E1095, Type I, Class B requirements
With heavy uniform walls and strong, fire-polished rims and stems to provide a long service life.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.