Funnels with long stem, KIMBLE®
Supplier: KIMBLEKONTES
28980-150
KIML28980-150EA 256 SGD
KIML28980-150
Funnels with long stem, KIMBLE®
Funnels Filtration Funnels Filter Funnels
KIMAX®, Borosilicate glass, with 60° cone angle.
  • High resistance to chemical attack
  • Resistant to mechanical and thermal shock
  • ASTM Specification E1095, Type I, Class B requirements
With heavy uniform walls and strong, fire-polished rims and stems to provide a long service life.