Separating funnels, Squibb, [ST] glass stopcock and stopper, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
29048-250 29048-500
KIML29048-250EA 987 SGD
KIML29048-250 KIML29048-500
Separating funnels, Squibb, [ST] glass stopcock and stopper, KIMAX®, KIMBLE®
Funnels Separating Funnels
Manufactured from 33 expansion, low extractable borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
KIMAX® separating funnels with a [ST] ground glass stopcock and stopper.
Funnels come with lower stems having large internal diameter that allow complete separation and eliminates the necessity of emptying the funnel.
Certifications: Designed from ASTM Specification E1096, Type IV requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements and then RAY-SORB® processed.