Separating funnels, Squibb, glass stopper, PTFE stopcock, KIMBLE®
Supplier: KIMBLEKONTES
29048F-125
KIML29048F-125EA 910 SGD
KIML29048F-125
Separating funnels, Squibb, glass stopper, PTFE stopcock, KIMBLE®
Funnels Separating Funnels
Borosilicate glass, KIMAX®, with glass stopper and PTFE stopcock.
Funnels come with lower stems having large internal diameter that allow complete separation and eliminates the necessity of emptying the funnel.
Certifications: Designed from ASTM specification E1096, type IV requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
SPECIFICATIONS
Stem length 60 mm
Environmentally Preferable