Gas sampling tubes, KIMBLE®
Supplier: KIMBLEKONTES
30040-500
KIML30040-500EA 891 SGD
KIML30040-500
Gas sampling tubes, KIMBLE®
Tubes Gas Sampling Tubes
Manufactured from 33 expansion, low extractable borosilicate glass, KIMAX®.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.