Serological pipettes, wide tip, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
37034B-10
KIML37034B-10EA 420 SGD
KIML37034B-10
Serological pipettes, wide tip, KIMAX®, KIMBLE®
Pipettes Serological Pipettes
Borosilicate glass, reusable.
  • Wide tip opening for fast delivery
  • Constricted top for cotton-plugging
  • Colour-coded top
  • Graduated (0 to 9,5 ml)
These KIMAX® pipettes are intended for use with viscous liquids, slurries and suspensions.
Certifications: USP Type I and ASTM E438, Type I, Class B