Allihn condensers with full length water cooled joints, KIMBLE®
Supplier: KIMBLEKONTES
431500-2430
KIML431500-2430EA 412 SGD
KIML431500-2430
Allihn condensers with full length water cooled joints, KIMBLE®
Condensers
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use in refluxing operations, including soxhlet extractions. It has a straight outlet tube at the top and a water-cooled standard taper inner drip joint at the bottom.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.