Culture tubes, round bottom, reusable, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
45060-25150 45060-16150 45060-650
KIML45060-25150EA 746 SGD
KIML45060-25150 KIML45060-16150 KIML45060-650
Culture tubes, round bottom, reusable, KIMAX®, KIMBLE®
Tubes Culture Tubes
Manufactured from 51 expansion borosilicate glass.
  • No marking spot
  • The 6×50 mm size is often referred to as a 'Durham tube'
Tubes with fire-polished top rims and uniform wall thicknesses for maximum heat transfer and chemical resistance.
Certifications: Designed from ASTM Specification E982, Type IV requirements. Glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.