Centrifuge tubes, graduated, beaded rim, KIMBLE®
Supplier: KIMBLEKONTES
45164-15
KIML45164-15EA 417 SGD
KIML45164-15
Centrifuge tubes, graduated, beaded rim, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass, KIMAX®, graduated.
  • Calibrated to contain
  • Marking spot is sand-blasted on both sizes
  • Graduated with permanent red stain
Standard tapered reusable tube has a beaded top for added strength.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.