Centrifuge tubes, graduated, beaded rim, KIMBLE®
Supplier: KIMBLEKONTES
45165-15
KIML45165-15EA 417 SGD
KIML45165-15
Centrifuge tubes, graduated, beaded rim, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass, KIMAX®, graduated.
  • Calibrated to contain
  • Graduated with white ceramic enamel
These standard tapered tube is reusable, and its top beaded for strength.
Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.