Centrifuge tubes, conical bottom, beaded rim, graduated, KIMBLE®
Supplier: KIMBLEKONTES
45167-50
KIML45167-50EA 683 SGD
KIML45167-50
Centrifuge tubes, conical bottom, beaded rim, graduated, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass, KIMAX®, graduated, beaded.
  • Calibrated to contain
  • Graduation in durable black ceramic enamel
Used in the determination of the solubility index of dry milk solids or where products as specified by the American Dairy Products Institute. Also used in pulp determination of citrus products.
Tubes are graduated upward to 1,0 ml in divisions of 0,1 ml; from 1 to 2 ml in divisions of 0,2 ml; from 2 to 10 ml in divisions of 0,5 ml; from 10 to 20 ml in divisions of 1 ml.
Certifications: Glass conforms to USP Type I and ASTM E438, TypeMore Product Information