Centrifuge tube, conical bottom, graduated, with PTFE stopper, KIMBLE®
Supplier: KIMBLEKONTES
45176-50
KIML45176-50EA 649 SGD
KIML45176-50
Centrifuge tube, conical bottom, graduated, with PTFE stopper, KIMBLE®
Tubes Centrifuge Tubes
Borosilicate glass KIMAX®, graduated, with PTFE stopper.
  • Calibrated to contain
  • Scale and legend are printed in permanent brown stain
  • With sandblasted marking spot
Certifications: Glass conforms USP Type I and ASTM E438, Type I, class A requirements.
Caution: Do not centrifuge with stopper.