Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Supplier: KIMBLEKONTES
45212-50 45212-35
KIML45212-50EA 496 SGD
KIML45212-50 KIML45212-35
Centrifuge tube, round bottom, with screw cap, KIMAX®, KIMBLE®
Tubes Centrifuge Tubes
Tube has a heavy wall to withstand higher speeds in centrifuging.
It is especially useful in separating neutralised sediment for culturing from sputum specimens, as well as many other clinical and microbiological applications.
Delivery information: Supplied with unattached autoclavable cap with cemented-in rubber liner.